Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APM1
Entrez Gene ID
855869
Gene Name
Gene Aliases
YAP54, YPL259C
Chromosome Location
Chr.XVI: 51244 - 52671 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184073.1 NP_015064.1 1 - 1 1250 64

View Full Product Details