Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMK1
Entrez Gene ID
856167
Gene Name
Gene Aliases
YPR054W
Chromosome Location
Chr.XVI: 666280 - 667446 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184151.1 NP_015379.1 1 - 1 1046 90

View Full Product Details