Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRL1
Entrez Gene ID
856194
Gene Name
Gene Aliases
YPR079W
Chromosome Location
Chr.XVI: 698869 - 700014 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184176.1 NP_015404.1 1 - 1 1049 107

View Full Product Details