Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BET2
Entrez Gene ID
856306
Gene Name
Gene Aliases
YPR176C
Chromosome Location
Chr.XVI: 891101 - 892078 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184273.1 NP_015502.1 1 - 1 856 69

View Full Product Details