Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD14
Entrez Gene ID
855241
Gene Name
Gene Aliases
YMR201C
Chromosome Location
Chr.XIII: 665845 - 667044 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182708.1 NP_013928.1 2 - 2 1054 78

View Full Product Details