Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP13
Entrez Gene ID
397346
Gene Name
matrix metallopeptidase 13
Gene Aliases
MMP-13
Chromosome Location
Chr.9: 33549003 - 33616700 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003129808.5 XP_003129856.2 - 748 69
XM_021062710.1 XP_020918369.1 - 1396 69
XM_021062711.1 XP_020918370.1 - 1920 69
XM_021062712.1 XP_020918371.1 - 1517 69
GenBank mRNA AF069643.1 - 1 - 2 50 69

View Full Product Details