Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
KDM5C
Entrez Gene ID
100037295
Gene Name
lysine demethylase 5C
Gene Aliases
JARID1C, SMCX
Chromosome Location
Chr.X: 45968812 - 46003215 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF132069.1 - 2 - 2 145 78

View Full Product Details