Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
MRPL32
Entrez Gene ID
396626
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
-
Chromosome Location
Chr.18: 51865077 - 51909291 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003360208.4 XP_003360256.3 - 362 85
GenBank mRNA AK343710.1 - 3 - 3 341 85
AK348381.1 - 3 - 3 342 85
AK389729.1 - 3 - 3 350 85
AY439280.1 - 2 - 2 53 85
AY609957.1 - 3 - 3 339 85

View Full Product Details