Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLN3
Entrez Gene ID
503836
Gene Name
relaxin 3
Gene Aliases
insl7
Chromosome Location
Chr.2: 65149883 - 65173306 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045679.1 NP_001039144.1 2 - 2 356 110
XM_005661236.3 XP_005661293.1 - 827 110
XM_013994666.2 XP_013850120.1 - 693 110
XM_021081988.1 XP_020937647.1 - 798 110
XM_021081995.1 XP_020937654.1 - 748 110

View Full Product Details