Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RGN
Entrez Gene ID
768107
Gene Name
regucalcin
Gene Aliases
GNL, SMP-30
Chromosome Location
Chr.X: 41725660 - 41749736 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077220.1 NP_001070688.1 5 - 6 854 62
XM_005673573.3 XP_005673630.1 - 1071 62
XM_005673574.3 XP_005673631.1 - 1028 62
XM_005673575.3 XP_005673632.1 - 1121 62
XM_005673576.3 XP_005673633.1 - 1194 62
GenBank mRNA AK233038.1 - 6 - 7 971 62
AK233394.1 - 6 - 7 1187 62
AK394337.1 - 6 - 7 976 62
DQ972961.1 - 5 - 6 854 62

View Full Product Details