Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
CD80
Entrez Gene ID
397161
Gene Name
CD80 molecule
Gene Aliases
B7-1
Chromosome Location
Chr.13: 140693312 - 140730541 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214087.1 NP_999252.1 5 - 5 1047 99
XM_021068533.1 XP_020924192.1 - 1166 99
XM_021068534.1 XP_020924193.1 - 1164 96
XM_021068535.1 XP_020924194.1 - 1165 96
GenBank mRNA AB049760.1 - 5 - 5 1047 99
AF203443.1 - 5 - 5 1032 99
AF455811.1 - 4 - 4 725 96

View Full Product Details