Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CCND2
Entrez Gene ID
397162
Gene Name
cyclin D2
Gene Aliases
-
Chromosome Location
Chr.5: 66092483 - 66114575 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214088.1 NP_999253.1 5 - 5 1053 97
GenBank mRNA AB046174.1 - 5 - 5 1053 97
AK234273.1 - 5 - 5 1093 97

View Full Product Details