The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214126.1 | NP_999291.1 | 3 - 4 | 347 | 99 |
GenBank mRNA | AB040443.1 | - | 3 - 4 | 347 | 99 |
AF263915.1 | - | 3 - 4 | 217 | 99 | |
HQ110108.1 | - | 3 - 4 | 347 | 99 |