Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX1
Entrez Gene ID
397403
Gene Name
glutathione peroxidase 1
Gene Aliases
-
Chromosome Location
Chr.13: 31916269 - 31917337 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214201.1 NP_999366.1 2 - 2 350 60
GenBank mRNA AF532927.1 - 2 - 2 350 60
AJ010340.1 - 1 - 1 107 60
AK231261.1 - 2 - 2 442 60
AK239914.1 - 2 - 2 427 60
AY743601.1 - 1 - 1 102 60
KJ686126.1 - 2 - 2 377 60

View Full Product Details