Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
MYOT
Entrez Gene ID
100101550
Gene Name
myotilin
Gene Aliases
TTID
Chromosome Location
Chr.2: 139899245 - 139919095 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099941.2 NP_001093411.1 9 - 9 1544 128
GenBank mRNA DQ157551.2 - 9 - 9 1545 128

View Full Product Details