Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4A3
Entrez Gene ID
100101926
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
DDX48, eIF4A-III
Chromosome Location
Chr.12: 2285679 - 2311750 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100193.1 NP_001093663.1 8 - 9 888 94
GenBank mRNA DQ351286.1 - 8 - 9 888 94

View Full Product Details