Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNCG
Entrez Gene ID
100125343
Gene Name
synuclein gamma
Gene Aliases
-
Chromosome Location
Chr.14: 87916353 - 87922111 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103211.1 NP_001096681.1 1 - 2 162 72
XM_013983396.2 XP_013838850.1 - 586 72
XM_013983397.2 XP_013838851.1 - 294 72
GenBank mRNA AK236982.1 - 1 - 2 219 72
AK348791.1 - 1 - 2 233 72
AK348968.1 - 1 - 2 235 72
AK391883.1 - 1 - 2 236 72
EF104640.1 - 1 - 2 162 72

View Full Product Details