Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNCG
Entrez Gene ID
100125343
Gene Name
synuclein gamma
Gene Aliases
-
Chromosome Location
Chr.14: 87916353 - 87922111 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103211.1 NP_001096681.1 4 - 5 407 65
XM_013983396.2 XP_013838850.1 - 831 65
XM_013983397.2 XP_013838851.1 - 539 65
GenBank mRNA AK236982.1 - 4 - 5 464 65
AK348791.1 - 4 - 5 478 65
AK348968.1 - 4 - 5 480 65
AK391883.1 - 4 - 5 481 65
EF104640.1 - 4 - 5 407 65

View Full Product Details