Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPTL1
Entrez Gene ID
100126165
Gene Name
angiopoietin like 1
Gene Aliases
-
Chromosome Location
Chr.9: 120656313 - 120684313 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109947.1 NP_001103417.1 3 - 4 1289 90
XM_005667787.3 XP_005667844.1 - 90
GenBank mRNA EF154830.1 - 3 - 4 1289 90

View Full Product Details