Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNPY3
Entrez Gene ID
100126290
Gene Name
canopy FGF signaling regulator 3
Gene Aliases
TNRC5
Chromosome Location
Chr.7: 38014766 - 38025752 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110428.1 NP_001103898.1 1 - 2 300 64
XM_021097866.1 XP_020953525.1 - 316 64
GenBank mRNA AK234034.1 - 1 - 2 238 64
AK234305.1 - 1 - 2 274 64
AK238459.1 - 1 - 2 230 64
AY610363.1 - 1 - 2 180 64

View Full Product Details