Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASS4
Entrez Gene ID
100126293
Gene Name
Cas scaffold protein family member 4
Gene Aliases
C17H20orf32, C20orf32
Chromosome Location
Chr.17: 56990774 - 57032306 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110431.2 NP_001103901.2 3 - 4 692 86

View Full Product Details