Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C4A
Entrez Gene ID
445467
Gene Name
complement C4A (Rodgers blood group)
Gene Aliases
C4
Chromosome Location
Chr.7: 24068019 - 24083060 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123089.2 NP_001116561.1 16 - 17 2122 67
GenBank mRNA EU076594.1 - 16 - 17 2123 67

View Full Product Details