Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOK5
Entrez Gene ID
100144540
Gene Name
docking protein 5
Gene Aliases
-
Chromosome Location
Chr.17: 55391074 - 55541561 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123107.1 NP_001116579.1 4 - 5 413 138
GenBank mRNA AK349342.1 - 4 - 5 671 138

View Full Product Details