Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PNMT
Entrez Gene ID
100144479
Gene Name
phenylethanolamine N-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.12: 22648396 - 22651520 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123164.1 NP_001116636.1 2 - 3 410 57
XM_021066037.1 XP_020921696.1 - 510 57
GenBank mRNA DQ917626.1 - 2 - 3 410 57

View Full Product Details