Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLAU
Entrez Gene ID
396985
Gene Name
plasminogen activator, urokinase
Gene Aliases
uPA
Chromosome Location
Chr.14: 76629313 - 76635173 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213945.1 NP_999110.1 3 - 4 196 74
GenBank mRNA AK391242.1 - 3 - 4 211 74
X02724.1 - - 196 74

View Full Product Details