The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038006.1 | NP_001033095.1 | 7 - 8 | 1690 | 79 |
XM_005662947.3 | XP_005663004.1 | - | 2315 | 79 | |
XM_013996572.2 | XP_013852026.1 | - | 2086 | 79 | |
XM_013996573.2 | XP_013852027.1 | - | 2174 | 79 | |
XM_013996574.2 | XP_013852028.1 | - | 2185 | 79 | |
GenBank mRNA | DQ091176.1 | - | 7 - 8 | 1690 | 79 |