Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTX1
Entrez Gene ID
664654
Gene Name
metaxin 1
Gene Aliases
Metaxin-1
Chromosome Location
Chr.4: 94606576 - 94611439 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039750.1 NP_001034839.1 4 - 5 339 73
GenBank mRNA AB239841.1 - 4 - 5 350 73
AK234275.1 - 4 - 5 350 73
AK347776.1 - 4 - 5 326 73
AK392795.1 - 4 - 5 343 73
DQ402035.1 - 4 - 5 339 73

View Full Product Details