Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APOM
Entrez Gene ID
692188
Gene Name
apolipoprotein M
Gene Aliases
-
Chromosome Location
Chr.7: 23768974 - 23773324 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040640.1 NP_001035730.1 2 - 3 316 76
XM_021098346.1 XP_020954005.1 - 2187 76
GenBank mRNA AY610077.1 - 2 - 3 349 76
DQ329240.2 - 2 - 3 316 76

View Full Product Details