The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_213803.1 | NP_998968.1 | 3 - 4 | 348 | 65 |
XM_005661643.3 | XP_005661700.1 | - | 869 | 65 | |
XM_021080318.1 | XP_020935977.1 | - | 1352 | 65 | |
XM_021080328.1 | XP_020935987.1 | - | 1349 | 65 | |
GenBank mRNA | AF385626.2 | - | 3 - 4 | 348 | 65 |
EU682383.1 | - | 3 - 4 | 345 | 65 |