Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA7
Entrez Gene ID
397125
Gene Name
serpin family A member 7
Gene Aliases
tbg
Chromosome Location
Chr.X: 86708624 - 86714324 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214058.2 NP_999223.1 3 - 4 938 71
XM_021079495.1 XP_020935154.1 - 957 71
GenBank mRNA AF204929.1 - 3 - 4 939 71

View Full Product Details