Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA7
Entrez Gene ID
397125
Gene Name
serpin family A member 7
Gene Aliases
tbg
Chromosome Location
Chr.X: 86708624 - 86714324 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214058.2 NP_999223.1 4 - 5 1092 79
XM_021079495.1 XP_020935154.1 - 1111 79
GenBank mRNA AF204929.1 - 4 - 5 1093 79

View Full Product Details