Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OVGP1
Entrez Gene ID
397140
Gene Name
oviductal glycoprotein 1
Gene Aliases
-
Chromosome Location
Chr.4: 108929034 - 108941040 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214070.1 NP_999235.1 2 - 3 69 64
GenBank mRNA AK347380.1 - 2 - 3 85 64
U43490.1 - 2 - 3 69 64

View Full Product Details