Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OVGP1
Entrez Gene ID
397140
Gene Name
oviductal glycoprotein 1
Gene Aliases
-
Chromosome Location
Chr.4: 108929034 - 108941040 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214070.1 NP_999235.1 8 - 9 918 88
GenBank mRNA AK347380.1 - 9 - 10 1024 88
U43490.1 - 8 - 9 918 88

View Full Product Details