Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU1F1
Entrez Gene ID
397325
Gene Name
POU class 1 homeobox 1
Gene Aliases
GHF-1, PIT-1, PIT-I, Pit1
Chromosome Location
Chr.13: 168510671 - 168530394 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214163.1 NP_999328.1 3 - 4 558 134
XM_005657130.3 XP_005657187.1 - 741 134
XM_021068539.1 XP_020924198.1 - 732 134
GenBank mRNA AF016251.2 - 3 - 4 558 134
HM163575.1 - 3 - 4 447 134
Y12475.1 - - 423 134

View Full Product Details