Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OAS1
Entrez Gene ID
397570
Gene Name
2'-5'-oligoadenylate synthetase 1
Gene Aliases
-
Chromosome Location
Chr.14: 38851083 - 38867427 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_214303.2 NP_999468.2 2 - 3 537 83
XM_021073680.1 XP_020929339.1 - 661 83
GenBank mRNA AJ225090.1 - 2 - 3 510 83
AK230597.1 - 2 - 3 529 83
AK230683.1 - 2 - 3 541 83
AK234442.1 - 2 - 3 612 83
AY550259.1 - 2 - 3 537 83

View Full Product Details