Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
MCEE
Entrez Gene ID
100037986
Gene Name
methylmalonyl-CoA epimerase
Gene Aliases
-
Chromosome Location
Chr.1: 143993714 - 144022372 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001097471.2 NP_001090940.1 2 - 2 349 80
XM_021072602.1 XP_020928261.1 - 436 80
GenBank mRNA AY610420.1 - 2 - 2 368 80
DQ629149.1 - 2 - 2 351 80

View Full Product Details