Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM21
Entrez Gene ID
100302538
Gene Name
tripartite motif containing 21
Gene Aliases
-
Chromosome Location
Chr.9: 5782133 - 5790628 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163649.2 NP_001157121.1 2 - 3 535 61
XM_005656620.3 XP_005656677.2 - 694 61
XM_005656621.3 XP_005656678.2 - 754 61
GenBank mRNA AK234261.1 - 3 - 4 594 61
AK349885.1 - 3 - 4 668 61
AK393012.1 - 3 - 4 591 61
GQ245762.1 - 2 - 3 537 61

View Full Product Details