Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTUB1
Entrez Gene ID
100302092
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
Gene Aliases
-
Chromosome Location
Chr.2: 8088528 - 8095555 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162403.1 NP_001155875.1 1 - 2 83 93
GenBank mRNA AK232154.1 - 1 - 2 80 93
AK392248.1 - 1 - 2 95 93
AK398824.1 - 1 - 2 70 93
AY609773.1 - 1 - 2 77 93
EU310930.1 - 1 - 2 83 93

View Full Product Details