Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SPATA17
Entrez Gene ID
100415922
Gene Name
spermatogenesis associated 17
Gene Aliases
-
Chromosome Location
Chr.10: 7797448 - 8120892 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184753.1 NP_001171682.1 6 - 7 518 73
XM_021063977.1 XP_020919636.1 - 584 73
GenBank mRNA GU475009.1 - 6 - 7 518 73

View Full Product Details