Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTFR2
Entrez Gene ID
100157026
Gene Name
mitochondrial fission regulator 2
Gene Aliases
FAM54A
Chromosome Location
Chr.1: 27906162 - 27998861 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190176.1 NP_001177105.1 3 - 4 546 124
XM_005654380.3 XP_005654437.1 - 3173 124
XM_013984860.2 XP_013840314.1 - 3173 124
GenBank mRNA AK232190.1 - 3 - 4 546 124
AK345725.1 - 3 - 4 546 124

View Full Product Details