Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTI
Entrez Gene ID
100156830
Gene Name
pancreatic trypsin inhibitor
Gene Aliases
-
Chromosome Location
Chr.17: 47773060 - 47780746 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190215.1 NP_001177144.1 3 - 4 569 110
GenBank mRNA AK238090.1 - 3 - 4 570 110
AK350831.1 - 3 - 4 571 110

View Full Product Details