Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMIM19
Entrez Gene ID
100152445
Gene Name
small integral membrane protein 19
Gene Aliases
C17H8orf40, C8orf40
Chromosome Location
Chr.17: 11549907 - 11566898 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190222.1 NP_001177151.1 2 - 3 398 69
NM_001190223.1 NP_001177152.1 2 - 3 364 69
NM_001190224.1 NP_001177153.1 3 - 4 474 69
GenBank mRNA AK232872.1 - 2 - 3 328 69
AK233265.1 - 3 - 4 474 69
AK233963.1 - 2 - 3 241 69
AK236795.1 - 2 - 3 383 69
AK238663.1 - 2 - 3 235 69
AK346585.1 - 2 - 3 164 69
AK389403.1 - 2 - 3 398 69
AK394516.1 - 2 - 3 224 69
AK397522.1 - 2 - 3 364 69
AK398631.1 - 2 - 3 224 69
AK398690.1 - 2 - 3 348 69
AK398918.1 - 2 - 3 259 69

View Full Product Details