Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ODF3
Entrez Gene ID
100499231
Gene Name
outer dense fiber of sperm tails 3
Gene Aliases
-
Chromosome Location
Chr.2: 20869 - 25845 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348539.1 NP_001335468.1 6 - 7 805 73
XM_021077666.1 XP_020933325.1 - 2014 73
GenBank mRNA HM138511.1 - 6 - 7 806 73

View Full Product Details