Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGO1
Entrez Gene ID
100499510
Gene Name
argonaute RISC component 1
Gene Aliases
EIF2C1
Chromosome Location
Chr.6: 92038104 - 92076477 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194976.1 NP_001181905.1 9 - 10 1141 67
XM_005665209.3 XP_005665266.1 - 1253 67
XM_021093293.1 XP_020948952.1 - 843 67
GenBank mRNA AK344581.1 - 8 - 9 1041 67
GQ329714.1 - 9 - 10 1141 67

View Full Product Details