Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CMYA5
Entrez Gene ID
100505410
Gene Name
cardiomyopathy associated 5
Gene Aliases
-
Chromosome Location
Chr.2: 88391104 - 88506336 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197307.1 NP_001184236.1 8 - 9 11656 100
XM_021077698.1 XP_020933357.1 - 11711 100
GenBank mRNA AK394690.1 - 6 - 7 720 100
FJ208850.1 - 8 - 9 11656 100

View Full Product Details