Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PALM2AKAP2
Entrez Gene ID
100152369
Gene Name
PALM2-AKAP2 fusion
Gene Aliases
AKAP2, PALM2, PALM2-AKAP2
Chromosome Location
Chr.1: 250824055 - 250968545 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204195.2 NP_001191124.2 5 - 6 654 95
NM_001370837.1 NP_001357766.1 5 - 6 654 95
GenBank mRNA AK234430.1 - 5 - 6 683 95
AK347588.1 - 5 - 6 654 95

View Full Product Details