Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P2RY11
Entrez Gene ID
100533477
Gene Name
purinergic receptor P2Y11
Gene Aliases
-
Chromosome Location
Chr.2: 68962996 - 68966224 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204436.1 NP_001191365.1 1 - 2 38 60
GenBank mRNA AK231475.1 - 1 - 2 38 60
AK395789.1 - 1 - 2 52 60

View Full Product Details