Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMAP-23
Entrez Gene ID
100170138
Gene Name
antibacterial protein
Gene Aliases
PMAP23, PR-39
Chromosome Location
Chr.13: 30959802 - 30961519 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129976.1 NP_001123448.1 3 - 4 387 82
GenBank mRNA L26053.1 - 3 - 4 387 82

View Full Product Details