Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN19
Entrez Gene ID
100294682
Gene Name
claudin 19
Gene Aliases
-
Chromosome Location
Chr.6: 168695246 - 168699683 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160084.1 NP_001153556.1 4 - 5 653 95
GenBank mRNA FJ875104.1 - 4 - 5 653 95

View Full Product Details