Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACTC1
Entrez Gene ID
100152267
Gene Name
actin alpha cardiac muscle 1
Gene Aliases
-
Chromosome Location
Chr.1: 136281119 - 136286551 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170517.2 NP_001163988.1 1 - 2 230 129

View Full Product Details